siRNA Details
| Gene Symbol: PLK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005030.5 | NP_005021.2 | 5 | 1049 | ||
| GenBank mRNA | AB209179.1 | - | 4 | 2908 | ||
| AK303263.1 | - | 6 | 665 | |||
| AK308276.1 | - | 6 | 906 | |||
| AK313227.1 | - | 5 | 973 | |||
| BC002369.2 | - | 5 | 941 | 2822226 | ||
| BC003002.1 | - | 5 | 919 | 2823623 | ||
| BC014846.1 | - | 5 | 894 | 3854860 | ||
| U01038.1 | - | 5 | 951 | |||
| X73458.1 | - | 5 | 965 | |||
| X75932.1 | - | 5 | 951 | |||

PLK1
Genomic Map