siRNA Details
| Gene Symbol: PNN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002687.3 | NP_002678.2 | 9 | 1127 | ||
| GenBank mRNA | AK223612.1 | - | 9 | 1062 | ||
| AK300164.1 | - | 2 | 376 | |||
| AK303136.1 | - | 5 | 718 | |||
| AK304622.1 | - | 1 | 232 | |||
| BC062602.1 | - | 9 | 1038 | 5491466 | ||
| Y09703.1 | - | 9 | 1037 | |||

PNN
Genomic Map