siRNA Details
| Gene Symbol: PNN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002687.3 | NP_002678.2 | 9 | 1602 | ||
| GenBank mRNA | AK223612.1 | - | 9 | 1537 | ||
| AK292579.1 | - | 9 | 1586 | |||
| AK300164.1 | - | 2 | 851 | |||
| AK303136.1 | - | 5 | 1193 | |||
| AK304622.1 | - | 1 | 707 | |||
| BC062602.1 | - | 9 | 1513 | 5491466 | ||
| BM850066.1 | - | 1 | 79 | |||
| Y09703.1 | - | 9 | 1512 | |||

PNN
Genomic Map