siRNA Details
| Gene Symbol: PNN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002687.3 | NP_002678.2 | 6 | 545 | ||
| GenBank mRNA | AK223612.1 | - | 6 | 480 | ||
| AK292579.1 | - | 6 | 529 | |||
| AK303136.1 | - | 2 | 136 | |||
| BC062602.1 | - | 6 | 456 | 5491466 | ||
| Y09703.1 | - | 6 | 455 | |||

PNN
Genomic Map