siRNA Details
| Gene Symbol: UBL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007106.3 | NP_009037.1 | 4 | 1431 | ||
| GenBank mRNA | AF044221.1 | - | 4 | 390 | ||
| AK056580.1 | - | 4 | 1326 | |||
| AK307361.1 | - | 4 | 375 | |||
| AK309253.1 | - | 4 | 360 | |||
| AK311846.1 | - | 4 | 474 | |||
| AL080177.1 | - | 4 | 395 | |||
| BC044582.1 | - | 4 | 874 | 5262176 | ||
| BC059385.1 | - | 4 | 771 | 5287541 | ||
| CR533458.1 | - | 4 | 286 | |||

UBL3
Genomic Map