siRNA Details
| Gene Symbol: SEPT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009939.2 | NP_001009939.1 | 9 | 1180 | ||
| NM_002688.5 | NP_002679.2 | 10 | 986 | |||
| GenBank mRNA | AK056273.1 | - | 9 | 1157 | ||
| AK295940.1 | - | 10 | 855 | |||
| BC025261.1 | - | 10 | 950 | 4622405 | ||
| CR456545.1 | - | 10 | 866 | |||
| KJ534756.1 | - | 10 | 861 | |||
| U74628.1 | - | 10 | 948 | |||
| Y11593.1 | - | 10 | 906 | |||
| Gene Symbol: SEPT5-GP1BB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037611.1 | 9 | 2406 | |||
| NR_037612.1 | 9 | 910 | ||||
| GenBank mRNA | AB209168.1 | - | 9 | 2406 | ||
| BC024304.1 | - | 2 | 67 | |||
| U59632.1 | - | 9 | 910 | |||

SEPT5
Genomic Map