siRNA Details
| Gene Symbol: POLD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127218.2 | NP_001120690.1 | 8, 9 | 1298 | ||
| NM_001256879.1 | NP_001243808.1 | 9, 10 | 1659 | |||
| NM_006230.3 | NP_006221.2 | 8, 9 | 1125 | |||
| XM_011515431.1 | XP_011513733.1 | 8, 9 | 1121 | |||
| GenBank mRNA | AK292347.1 | - | 9, 10 | 1659 | ||
| AK298168.1 | - | 8, 9 | 989 | |||
| AK312294.1 | - | 8, 9 | 1258 | |||
| BC000459.2 | - | 8, 9 | 1088 | 2822169 | ||
| BC030001.2 | - | 7, 8 | 1065 | |||
| U21090.1 | - | 8, 9 | 1087 | |||

POLD2
Genomic Map