siRNA Details
| Gene Symbol: POU2F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198783.1 | NP_001185712.1 | 15 | 2162 | ||
| NM_001198786.1 | NP_001185715.1 | 14 | 1840 | |||
| NM_002697.3 | NP_002688.3 | 15 | 2029 | |||
| NR_037163.1 | 16 | 2073 | ||||
| XM_011509653.2 | XP_011507955.1 | 16 | 1981 | |||
| XM_011509654.2 | XP_011507956.1 | 15 | 1942 | |||
| XM_011509655.1 | XP_011507957.1 | 16 | 1981 | |||
| GenBank mRNA | AK091438.1 | - | 17 | 2457 | ||
| AK303201.1 | - | 14 | 1778 | |||
| AY113189.1 | - | 15 | 1981 | |||
| BC001664.2 | - | 15 | 1945 | 2966289 | ||
| BC003571.1 | - | 15 | 1945 | 3544378 | ||
| BC052274.1 | - | 16 | 2053 | 4622256 | ||
| KT438684.1 | - | 17 | 2140 | |||
| X13403.1 | - | 15 | 1950 | |||

POU2F1
Genomic Map