siRNA Details
Gene Symbol: POU4F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002700.2 | NP_002691.1 | 2 | 952 | ||
GenBank mRNA | BC104923.1 | - | 2 | 952 | 8143926 | |
BC112207.1 | - | 2 | 952 | 8327675 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: POU4F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002700.2 | NP_002691.1 | 2 | 952 | ||
GenBank mRNA | BC104923.1 | - | 2 | 952 | 8143926 | |
BC112207.1 | - | 2 | 952 | 8327675 |