siRNA Details
| Gene Symbol: PPP1R3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005398.5 | NP_005389.1 | 2 | 700 | ||
| GenBank mRNA | AF087874.1 | - | 2 | 642 | ||
| AK299392.1 | - | 3 | 541 | |||
| AK299398.1 | - | 3 | 446 | |||
| AK313149.1 | - | 2 | 670 | |||
| BC012625.1 | - | 2 | 625 | 4245774 | ||
| BX537399.1 | - | 2 | 701 | |||
| Y18207.1 | - | 2 | 659 | |||

PPP1R3C
Genomic Map