siRNA Details
Gene Symbol: PPP2R5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199756.1 | NP_001186685.1 | 2 | 356 | ||
NM_006243.3 | NP_006234.1 | 2 | 824 | |||
GenBank mRNA | AK302202.1 | - | 2 | 356 | ||
AK312530.1 | - | 3 | 639 | |||
BC022474.1 | - | 2 | 410 | 4815091 | ||
BC110883.1 | - | 2 | 533 | 6045358 | ||
BP326450.1 | - | 2 | 349 | |||
L42373.1 | - | 2 | 821 |
