siRNA Details
| Gene Symbol: PRKACG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002732.3 | NP_002723.2 | 1 | 946 | ||
| GenBank mRNA | BC039888.1 | - | 1 | 946 | 5165721 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRKACG| Gene Symbol: PRKACG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002732.3 | NP_002723.2 | 1 | 946 | ||
| GenBank mRNA | BC039888.1 | - | 1 | 946 | 5165721 | |