siRNA Details
| Gene Symbol: PRKCQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242413.2 | NP_001229342.1 | 12 | 1421 | ||
| NM_001282644.1 | NP_001269573.1 | 12 | 1419 | |||
| NM_001282645.1 | NP_001269574.1 | 11 | 1360 | |||
| NM_001323265.1 | NP_001310194.1 | 12 | 1476 | |||
| NM_001323266.1 | NP_001310195.1 | 11 | 1358 | |||
| NM_001323267.1 | NP_001310196.1 | 12 | 1419 | |||
| NM_006257.4 | NP_006248.1 | 12 | 1421 | |||
| XM_005252496.4 | XP_005252553.1 | 12 | 1521 | |||
| XM_005252497.4 | XP_005252554.1 | 12 | 1523 | |||
| XM_006717465.3 | XP_006717528.1 | 14 | 3916 | |||
| XM_017016410.1 | XP_016871899.1 | 13 | 3827 | |||
| XM_017016411.1 | XP_016871900.1 | 13 | 3855 | |||
| GenBank mRNA | AK293935.1 | - | 11 | 1298 | ||
| AK308099.1 | - | 12 | 1394 | |||
| AK312411.1 | - | 12 | 1371 | |||
| AY702977.1 | - | 11 | 1321 | |||
| BC101465.1 | - | 11 | 1340 | 8068971 | ||
| BC113359.1 | - | 11 | 1340 | 8322411 | ||
| BX647657.1 | - | 11 | 1506 | |||
| L01087.1 | - | 12 | 1415 | |||
| L07032.1 | - | 11 | 1342 | |||

PRKCQ
Genomic Map