siRNA Details
| Gene Symbol: MAPK13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002754.4 | NP_002745.1 | 2 | 265 | ||
| NR_072996.1 | 2 | 265 | ||||
| GenBank mRNA | AB209586.1 | - | 1 | 1223 | ||
| AB451221.1 | - | 3 | 166 | |||
| AB451344.1 | - | 3 | 166 | |||
| AF004709.1 | - | 2 | 214 | |||
| AF092535.1 | - | 2 | 187 | |||
| AF100546.1 | - | 2 | 168 | |||
| AK222922.1 | - | 2 | 157 | |||
| AK308018.1 | - | 2 | 246 | |||
| BC000433.2 | - | 2 | 207 | 2819932 | ||
| BC001641.2 | - | 2 | 207 | 2819932 | ||
| BC004428.1 | - | 2 | 207 | 2819932 | ||
| BC085196.1 | - | 2 | 235 | 6107307 | ||
| BM921729.1 | - | 2 | 264 | |||
| BT007221.1 | - | 2 | 166 | |||
| CR536490.1 | - | 2 | 166 | |||
| CR541687.1 | - | 2 | 166 | |||
| DB473379.1 | - | 2 | 229 | |||
| DR006449.1 | - | 2 | 269 | |||
| KU178281.1 | - | 2 | 166 | |||
| KU178282.1 | - | 2 | 166 | |||
| U93232.1 | - | 2 | 173 | |||
| Y10488.1 | - | 2 | 188 | |||

MAPK13
Genomic Map