siRNA Details
| Gene Symbol: PRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000948.5 | NP_000939.1 | 5 | 1087 | ||
| NM_001163558.2 | NP_001157030.1 | 6 | 759 | |||
| XM_011514753.2 | XP_011513055.1 | 6 | 806 | |||
| XM_011514754.2 | XP_011513056.1 | 7 | 819 | |||
| GenBank mRNA | BC015850.1 | - | 5 | 1090 | 4657724 | |
| BC088370.1 | - | 5 | 617 | 30393428 | ||
| CD106243.1 | - | 2 | 97 | |||
| DQ839492.1 | - | 6 | 475 | |||
| GQ305133.1 | - | 6 | 759 | |||
| V00566.1 | - | 5 | 572 | |||
| X54393.1 | - | 6 | 709 | |||

PRL
Genomic Map