siRNA Details
| Gene Symbol: PRPS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204402.1 | NP_001191331.1 | 3 | 528 | ||
| NM_002764.3 | NP_002755.1 | 6 | 936 | |||
| GenBank mRNA | AK223169.1 | - | 6 | 753 | ||
| AK297968.1 | - | 7 | 774 | |||
| AK307748.1 | - | 5 | 752 | |||
| AK312706.1 | - | 6 | 850 | |||
| AK316467.1 | - | 3 | 442 | |||
| BC001605.1 | - | 6 | 831 | 3542584 | ||
| D00860.1 | - | 6 | 853 | |||
| X15331.1 | - | 6 | 797 | |||

PRPS1
Genomic Map