siRNA Details
| Gene Symbol: PRRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000950.2 | NP_000941.1 | 5 | 811 | ||
| NM_001142395.1 | NP_001135867.1 | 4 | 720 | |||
| NM_001173489.1 | NP_001166960.1 | 5 | 830 | |||
| NM_001173490.1 | NP_001166961.1 | 4 | 739 | |||
| GenBank mRNA | AF009242.1 | - | 5 | 723 | ||
| AK312904.1 | - | 4 | 659 | |||
| BC030786.2 | - | 4 | 703 | |||
| BC060833.1 | - | 4 | 659 | 30348133 | ||

PRRG1
Genomic Map