siRNA Details
| Gene Symbol: KLK7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243126.1 | NP_001230055.1 | 2 | 305 | ||
| NM_005046.3 | NP_005037.1 | 2 | 305 | |||
| NM_139277.2 | NP_644806.1 | 2 | 135 | |||
| GenBank mRNA | AF411214.1 | - | 2 | 279 | ||
| AY646152.1 | - | 1 | 37 | |||
| DA243628.1 | - | 2 | 300 | |||

KLK7
Genomic Map