siRNA Details
| Gene Symbol: PRSS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002773.4 | NP_002764.1 | 1 | 319 | ||
| GenBank mRNA | AK296656.1 | - | 1 | 284 | ||
| AK301619.1 | - | 1 | 284 | |||
| AK309955.1 | - | 1 | 260 | |||
| AU142128.1 | - | 1 | 319 | |||
| BC001462.2 | - | 1 | 230 | 3138532 | ||
| L41351.1 | - | 1 | 283 | |||

PRSS8
Genomic Map