siRNA Details
Gene Symbol: PSKH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006742.2 | NP_006733.1 | 2 | 1058 | ||
GenBank mRNA | AJ272212.1 | - | 2 | 1018 | ||
AK309272.1 | - | 2 | 1058 | |||
BC062616.1 | - | 2 | 1029 | 5188431 | ||
M14504.1 | - | 1 | 294 |
