siRNA Details
| Gene Symbol: PSMD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270427.1 | NP_001257356.1 | 9 | 1267 | ||
| NM_005047.3 | NP_005038.1 | 10 | 1396 | |||
| XM_017014928.1 | XP_016870417.1 | 9 | 1196 | |||
| GenBank mRNA | AK026925.1 | - | 6 | 699 | ||
| AK291051.1 | - | 10 | 1297 | |||
| AK303007.1 | - | 9 | 1197 | |||
| BC014478.1 | - | 10 | 1285 | 4878087 | ||
| D31889.1 | - | 10 | 1283 | |||
| S79862.1 | - | 10 | 1287 | |||

PSMD5
Genomic Map