siRNA Details
| Gene Symbol: PSMD12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001316341.1 | NP_001303270.1 | 11 | 1354 | ||
| NM_002816.4 | NP_002807.1 | 9 | 1070 | |||
| NM_174871.3 | NP_777360.1 | 8 | 1010 | |||
| GenBank mRNA | AB003103.1 | - | 9 | 1005 | ||
| AK091198.1 | - | 8 | 988 | |||
| AK222679.1 | - | 9 | 987 | |||
| AK307821.1 | - | 9 | 1020 | |||
| BC019062.2 | - | 9 | 1001 | 4111477 | ||
| BC065826.1 | - | 11 | 1274 | 30386487 | ||
| KU178288.1 | - | 9 | 962 | |||
| KU178289.1 | - | 9 | 994 | |||

PSMD12
Genomic Map