siRNA Details
| Gene Symbol: PSMD12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001316341.1 | NP_001303270.1 | 13 | 1647 | ||
| NM_002816.4 | NP_002807.1 | 11 | 1363 | |||
| NM_174871.3 | NP_777360.1 | 10 | 1303 | |||
| GenBank mRNA | AB003103.1 | - | 11 | 1298 | ||
| AK091198.1 | - | 10 | 1281 | |||
| AK222679.1 | - | 11 | 1280 | |||
| AK307821.1 | - | 11 | 1313 | |||
| BC019062.2 | - | 11 | 1294 | 4111477 | ||
| BC065826.1 | - | 13 | 1567 | 30386487 | ||
| KU178288.1 | - | 11 | 1255 | |||
| KU178289.1 | - | 11 | 1287 | |||

PSMD12
Genomic Map