siRNA Details
| Gene Symbol: PSME1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281528.1 | NP_001268457.1 | 7 | 533 | ||
| NM_001281529.1 | NP_001268458.1 | 7 | 533 | |||
| NM_006263.3 | NP_006254.1 | 7 | 533 | |||
| NM_176783.2 | NP_788955.1 | 7 | 533 | |||
| GenBank mRNA | AK311490.1 | - | 5 | 734 | ||
| AK312211.1 | - | 7 | 468 | |||
| BC000352.2 | - | 7 | 519 | 2961448 | ||
| BC007503.2 | - | 7 | 519 | 2961448 | ||
| BM470561.1 | - | 7 | 469 | |||
| BM981616.1 | - | 5 | 495 | |||
| BQ060125.1 | - | 7 | 491 | |||
| BT019337.1 | - | 7 | 428 | |||
| BU192084.1 | - | 7 | 464 | |||
| BU731923.1 | - | 5 | 487 | |||
| BX248293.1 | - | 3 | 201 | |||
| CR456780.1 | - | 7 | 428 | |||
| CR541658.1 | - | 7 | 428 | |||
| DQ185047.1 | - | 5 | 363 | |||
| GQ472237.1 | - | 7 | 519 | |||
| L07633.1 | - | 7 | 520 | |||

PSME1
Genomic Map