siRNA Details
| Gene Symbol: TAS2R38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176817.4 | NP_789787.4 | 1 | 898 | ||
| GenBank mRNA | BC104933.1 | - | 1 | 898 | 8143936 | |
| BC104937.1 | - | 1 | 898 | 8143940 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TAS2R38| Gene Symbol: TAS2R38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176817.4 | NP_789787.4 | 1 | 898 | ||
| GenBank mRNA | BC104933.1 | - | 1 | 898 | 8143936 | |
| BC104937.1 | - | 1 | 898 | 8143940 | ||