siRNA Details
| Gene Symbol: RAP2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021033.6 | NP_066361.1 | 2 | 716 | ||
| GenBank mRNA | AF205602.1 | - | 2 | 483 | ||
| AF493914.1 | - | 2 | 466 | |||
| AK311202.1 | - | 2 | 225 | |||
| AK315139.1 | - | 3 | 512 | |||
| BC041333.2 | - | 2 | 717 | 5278625 | ||
| BC070031.1 | - | 2 | 474 | 4815530 | ||
| X12534.1 | - | 2 | 469 | |||

RAP2A
Genomic Map