siRNA Details
| Gene Symbol: RARB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000965.4 | NP_000956.2 | 7, 8 | 1623 | ||
| NM_001290216.2 | NP_001277145.1 | 10, 11 | 1666 | |||
| NM_001290217.1 | NP_001277146.1 | 7, 8 | 1081 | |||
| NM_001290266.1 | NP_001277195.1 | 7, 8 | 1266 | |||
| NM_001290276.1 | NP_001277205.1 | 7, 8 | 1623 | |||
| NM_001290277.1 | NP_001277206.1 | 7, 8 | 1485 | |||
| NM_001290300.1 | NP_001277229.1 | 7, 8 | 1087 | |||
| NM_016152.3 | NP_057236.1 | 7, 8 | 1266 | |||
| NR_110892.1 | 6, 7 | 1462 | ||||
| NR_110893.1 | 6, 7 | 1418 | ||||
| GenBank mRNA | AB307688.1 | - | 7, 8 | 1154 | ||
| AF157483.1 | - | 7, 8 | 1265 | |||
| AK292032.1 | - | 7, 8 | 1173 | |||
| AK299756.1 | - | 7, 8 | 1241 | |||
| AY501390.1 | - | 7, 8 | 1055 | |||
| BC060794.1 | - | 7, 8 | 1305 | 30341884 | ||
| CD014020.1 | - | 6, 7 | 965 | |||
| CD014021.1 | - | 6, 7 | 1009 | |||
| CD014022.1 | - | 7, 8 | 1032 | |||
| CR536558.1 | - | 5, 6 | 818 | |||
| HQ692863.1 | - | 5, 6 | 818 | |||
| HQ692864.1 | - | 7, 8 | 1154 | |||
| X07282.1 | - | 7, 8 | 1497 | |||
| Y00291.1 | - | 7, 8 | 1475 | |||

RARB
Genomic Map