siRNA Details
| Gene Symbol: RARB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000965.4 | NP_000956.2 | 3 | 793 | ||
| NM_001290216.2 | NP_001277145.1 | 6 | 836 | |||
| NM_001290217.1 | NP_001277146.1 | 3 | 251 | |||
| NM_001290266.1 | NP_001277195.1 | 3 | 436 | |||
| NM_001290276.1 | NP_001277205.1 | 3 | 793 | |||
| NM_001290277.1 | NP_001277206.1 | 3 | 793 | |||
| NM_001290300.1 | NP_001277229.1 | 3 | 257 | |||
| NM_016152.3 | NP_057236.1 | 3 | 436 | |||
| NR_110892.1 | 3 | 793 | ||||
| NR_110893.1 | 3 | 793 | ||||
| GenBank mRNA | AB307688.1 | - | 3 | 324 | ||
| AF157483.1 | - | 3 | 435 | |||
| AK292032.1 | - | 3 | 343 | |||
| AK299756.1 | - | 3 | 411 | |||
| AY501390.1 | - | 3 | 225 | |||
| BC050415.2 | - | 1 | 19 | |||
| BC060794.1 | - | 3 | 475 | 30341884 | ||
| CD014020.1 | - | 3 | 340 | |||
| CD014021.1 | - | 3 | 340 | |||
| CD014022.1 | - | 3 | 340 | |||
| DA882771.1 | - | 3 | 251 | |||
| DC376623.1 | - | 3 | 411 | |||
| DQ083391.1 | - | 3 | 257 | |||
| HQ692864.1 | - | 3 | 324 | |||
| X07282.1 | - | 3 | 667 | |||
| Y00291.1 | - | 3 | 645 | |||

RARB
Genomic Map