siRNA Details
| Gene Symbol: RDH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199771.1 | NP_001186700.1 | 2 | 476 | ||
| NM_002905.3 | NP_002896.2 | 2 | 471 | |||
| GenBank mRNA | AK293313.1 | - | 2 | 427 | ||
| AK307157.1 | - | 2 | 298 | |||
| BC028298.1 | - | 2 | 430 | 4942311 | ||
| BI765016.1 | - | 2 | 382 | |||
| BP381700.1 | - | 2 | 471 | |||
| U43559.1 | - | 2 | 364 | |||
| U89717.1 | - | 2 | 401 | |||

RDH5
Genomic Map