siRNA Details
| Gene Symbol: RP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203288.1 | NP_976033.1 | 5, 6 | 486 | ||
| XM_011515468.2 | XP_011513770.1 | 5, 6 | 962 | |||
| GenBank mRNA | BC025928.1 | - | 5, 6 | 486 | 4903338 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RP9| Gene Symbol: RP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203288.1 | NP_976033.1 | 5, 6 | 486 | ||
| XM_011515468.2 | XP_011513770.1 | 5, 6 | 962 | |||
| GenBank mRNA | BC025928.1 | - | 5, 6 | 486 | 4903338 | |