siRNA Details
| Gene Symbol: RPS21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024.3 | NP_001015.1 | 5, 6 | 323 | ||
| GenBank mRNA | AV758927.1 | - | 284 | |||
| BC021990.1 | - | 5, 6 | 288 | 4066330 | ||
| CF124568.1 | - | 5, 6 | 364 | |||
| CR542132.1 | - | 233 | ||||
| L04483.1 | - | 5, 6 | 271 | |||

RPS21
Genomic Map