siRNA Details
| Gene Symbol: RPS21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024.3 | NP_001015.1 | 2, 3 | 144 | ||
| GenBank mRNA | AV758927.1 | - | 105 | |||
| BC018140.1 | - | 1, 2 | 81 | 3903320 | ||
| BC021990.1 | - | 2, 3 | 109 | 4066330 | ||
| BM840443.1 | - | 2, 3 | 144 | |||
| CF124568.1 | - | 2, 3 | 185 | |||
| CR542132.1 | - | 1, 2 | 54 | |||
| L04483.1 | - | 2, 3 | 92 | |||
| X79563.2 | - | 2, 3 | 91 | |||

RPS21
Genomic Map