siRNA Details
| Gene Symbol: CLIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001247997.1 | NP_001234926.1 | 16, 17 | 3177 | ||
| NM_002956.2 | NP_002947.1 | 15, 16 | 3144 | |||
| NM_198240.1 | NP_937883.1 | 14, 15 | 3039 | |||
| XM_005253593.2 | XP_005253650.1 | 16, 17 | 3240 | |||
| XM_006719552.3 | XP_006719615.1 | 15, 16 | 3233 | |||
| XM_017019786.1 | XP_016875275.1 | 15, 16 | 3308 | |||
| XM_017019787.1 | XP_016875276.1 | 15, 16 | 3123 | |||
| XM_017019788.1 | XP_016875277.1 | 14, 15 | 3128 | |||
| XM_017019789.1 | XP_016875278.1 | 14, 15 | 3287 | |||
| XM_017019790.1 | XP_016875279.1 | 14, 15 | 3029 | |||
| XM_017019791.1 | XP_016875280.1 | 14, 15 | 3116 | |||
| XM_017019792.1 | XP_016875281.1 | 13, 14 | 3011 | |||
| GenBank mRNA | AB209115.1 | - | 11, 12 | 1897 | ||
| AK126990.1 | - | 5, 6 | 827 | |||
| BC114213.1 | - | 13, 14 | 2954 | 7961952 | ||
| BC117209.1 | - | 15, 16 | 3064 | 40125760 | ||
| BC126305.1 | - | 15, 16 | 3064 | 8992021 | ||
| M97501.1 | - | 14, 15 | 3039 | |||
| X64838.1 | - | 15, 16 | 3121 | |||

CLIP1
Genomic Map