siRNA Details
| Gene Symbol: SDHB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003000.2 | NP_002991.2 | 2 | 272 | ||
| GenBank mRNA | AK312056.1 | - | 2 | 134 | ||
| BC007840.2 | - | 2 | 272 | 4124737 | ||
| BF796275.1 | - | 2 | 120 | |||
| D10245.1 | - | 1 | 67 | |||
| DQ403007.1 | - | 2 | 94 | |||
| M32246.1 | - | 1 | 67 | |||
| U17248.1 | - | 2 | 255 | |||

SDHB
Genomic Map