siRNA Details
| Gene Symbol: SEL1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005065.5 | NP_005056.3 | 14, 15 | 1500 | ||
| XM_005267988.2 | XP_005268045.1 | 14, 15 | 1383 | |||
| XM_005267989.3 | XP_005268046.1 | 13, 14 | 1434 | |||
| GenBank mRNA | AB020335.1 | - | 14, 15 | 1428 | ||
| AF052059.1 | - | 14, 15 | 1476 | |||
| AK075511.1 | - | 14, 15 | 1479 | |||

SEL1L
Genomic Map