siRNA Details
| Gene Symbol: SFRP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003014.3 | NP_003005.2 | 1 | 656 | ||
| GenBank mRNA | AF026692.1 | - | 1 | 527 | ||
| AK302357.1 | - | 1 | 578 | |||
| AK303977.1 | - | 2 | 476 | |||
| BC047684.1 | - | 1 | 514 | 5742936 | ||
| BC058911.1 | - | 1 | 534 | 5228231 | ||
| BT019679.1 | - | 1 | 270 | |||
| CR541731.1 | - | 1 | 270 | |||
| CR541755.1 | - | 1 | 270 | |||
| DQ420628.1 | - | 1 | 289 | |||
More Information
Set Membership: |
|

SFRP4
Genomic Map