siRNA Details
Gene Symbol: SGCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000337.5 | NP_000328.2 | 3 | 673 | ||
NM_001128209.1 | NP_001121681.1 | 2 | 627 | |||
NM_172244.2 | NP_758447.1 | 3 | 673 | |||
XM_005265966.4 | XP_005266023.1 | 3 | 501 | |||
XM_005265967.2 | XP_005266024.1 | 3 | 265 | |||
XM_011534621.2 | XP_011532923.1 | 2 | 330 | |||
XM_017009723.1 | XP_016865212.1 | 4 | 439 | |||
XM_017009724.1 | XP_016865213.1 | 4 | 716 | |||
GenBank mRNA | AK292794.1 | - | 3 | 265 | ||
BC008431.1 | - | 5 | 572 | |||
BC020740.1 | - | 3 | 266 | 4702959 | ||
BQ434817.1 | - | 3 | 673 | |||
BX537948.1 | - | 2 | 638 | |||
DA561069.1 | - | 2 | 220 | |||
U58331.1 | - | 2 | 192 | |||
X95191.1 | - | 1 | 154 |
