siRNA Details
| Gene Symbol: SGSH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000199.3 | NP_000190.1 | 8 | 1281 | ||
| XM_005257582.3 | XP_005257639.1 | 8 | 1390 | |||
| XM_011525127.2 | XP_011523429.1 | 9 | 1269 | |||
| XM_017024952.1 | XP_016880441.1 | 8 | 2126 | |||
| GenBank mRNA | AB209900.1 | - | 5 | 4076 | ||
| AK095969.1 | - | 7 | 1135 | |||
| AK222890.1 | - | 8 | 1214 | |||
| AK291257.1 | - | 8 | 1214 | |||
| BC047318.1 | - | 8 | 1206 | 5226903 | ||
| U30894.1 | - | 8 | 1206 | |||

SGSH
Genomic Map