siRNA Details
| Gene Symbol: SKI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003036.3 | NP_003027.1 | 6 | 1957 | ||
| XM_005244775.3 | XP_005244832.1 | 6 | 1933 | |||
| XM_005244776.4 | XP_005244833.1 | 6 | 5076 | |||
| XM_017002128.1 | XP_016857617.1 | 6 | 1740 | |||
| GenBank mRNA | X15218.1 | - | 6 | 1957 | ||

SKI
Genomic Map