siRNA Details
| Gene Symbol: SLC5A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014227.2 | NP_055042.1 | 6 | 561 | ||
| XM_006724308.3 | XP_006724371.1 | 7 | 812 | |||
| XM_011530342.2 | XP_011528644.1 | 8 | 930 | |||
| XM_011530343.2 | XP_011528645.1 | 6 | 476 | |||
| XM_011530344.2 | XP_011528646.1 | 5 | 441 | |||
| XM_017028920.1 | XP_016884409.1 | 8 | 891 | |||
| GenBank mRNA | AJ133127.1 | - | 6 | 575 | ||
| U41897.1 | - | 4 | 344 | |||

SLC5A4
Genomic Map