siRNA Details
| Gene Symbol: SLC6A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134367.3 | NP_001127839.2 | 11 | 1585 | ||
| NM_003043.5 | NP_003034.2 | 11 | 1589 | |||
| NR_103507.2 | 10 | 1464 | ||||
| XM_006713307.2 | XP_006713370.1 | 10 | 1393 | |||
| XM_011534030.2 | XP_011532332.1 | 10 | 1346 | |||
| XM_011534031.2 | XP_011532333.1 | 12 | 1717 | |||
| GenBank mRNA | AB209172.1 | - | 10 | 1390 | ||
| AK303649.1 | - | 10 | 1464 | |||
| AK308417.1 | - | 9 | 1065 | |||
| BC137128.1 | - | 11 | 1536 | 9021125 | ||
| BC137129.1 | - | 11 | 1532 | 9021126 | ||
| U09220.1 | - | 9 | 1264 | |||
| U16120.1 | - | 12 | 1640 | |||
| Z18956.1 | - | 9 | 1283 | |||

SLC6A6
Genomic Map