siRNA Details
| Gene Symbol: SLC6A13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190997.2 | NP_001177926.1 | 2 | 119 | ||
| NM_001243392.1 | NP_001230321.1 | 2 | 119 | |||
| NM_016615.4 | NP_057699.2 | 2 | 119 | |||
| XM_011521012.2 | XP_011519314.1 | 2 | 176 | |||
| XM_011521013.1 | XP_011519315.1 | 2 | 82 | |||
| XM_017019843.1 | XP_016875332.1 | 2 | 82 | |||
| XM_017019844.1 | XP_016875333.1 | 2 | 82 | |||
| XM_017019846.1 | XP_016875335.1 | 2 | 82 | |||
| XM_017019847.1 | XP_016875336.1 | 2 | 82 | |||
| GenBank mRNA | AF462445.1 | - | 2 | 83 | ||
| AK295904.1 | - | 2 | 82 | |||
| AK296127.1 | - | 2 | 82 | |||
| AK296209.1 | - | 2 | 82 | |||
| AK313511.1 | - | 2 | 75 | |||
| BC020867.1 | - | 2 | 82 | |||
| BC022392.1 | - | 2 | 82 | 4612245 | ||
| DA374290.1 | - | 2 | 119 | |||
| U76343.2 | - | 1 | 48 | |||

SLC6A13
Genomic Map