siRNA Details
| Gene Symbol: SLC11A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000578.3 | NP_000569.3 | 4 | 717 | ||
| XM_005246793.3 | XP_005246850.1 | 3 | 837 | |||
| XM_005246794.3 | XP_005246851.1 | 4 | 980 | |||
| XM_006712709.3 | XP_006712772.1 | 3 | 837 | |||
| XM_006712710.3 | XP_006712773.1 | 2 | 714 | |||
| XM_006712711.3 | XP_006712774.1 | 2 | 714 | |||
| XM_011511684.2 | XP_011509986.1 | 4 | 980 | |||
| XM_011511685.2 | XP_011509987.1 | 3 | 837 | |||
| XM_017004765.1 | XP_016860254.1 | 3 | 857 | |||
| XM_017004766.1 | XP_016860255.1 | 4 | 948 | |||
| XM_017004767.1 | XP_016860256.1 | 4 | 544 | |||
| GenBank mRNA | AB209650.1 | - | 2 | 2431 | ||
| AK294707.1 | - | 4 | 458 | |||
| AK298667.1 | - | 4 | 471 | |||
| AK303398.1 | - | 4 | 441 | |||
| AK307954.1 | - | 4 | 717 | |||
| AK310914.1 | - | 4 | 473 | |||
| AK311405.1 | - | 3 | 324 | |||
| BC033754.1 | - | 3 | 305 | |||
| BC071165.1 | - | 4 | 437 | 5756382 | ||
| D38171.1 | - | 2 | 224 | |||
| D50402.1 | - | 4 | 552 | |||
| D50403.1 | - | 4 | 463 | |||
| L32185.1 | - | 4 | 453 | |||

SLC11A1
Genomic Map