siRNA Details
| Gene Symbol: SLC14A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128588.3 | NP_001122060.3 | 5 | 624 | ||
| NM_001146036.2 | NP_001139508.2 | 5 | 553 | |||
| NM_001146037.1 | NP_001139509.1 | 3 | 721 | |||
| NM_001308278.1 | NP_001295207.1 | 3 | 298 | |||
| NM_015865.6 | NP_056949.4 | 4 | 466 | |||
| XM_005258329.1 | XP_005258386.1 | 5 | 600 | |||
| XM_006722526.3 | XP_006722589.1 | 5 | 601 | |||
| XM_011526142.1 | XP_011524444.1 | 4 | 1381 | |||
| XM_011526144.1 | XP_011524446.1 | 5 | 600 | |||
| GenBank mRNA | AK123681.1 | - | 2 | 1525 | ||
| AK289608.1 | - | 5 | 567 | |||
| AK294129.1 | - | 3 | 237 | |||
| BC050539.1 | - | 4 | 392 | 5162971 | ||
| DA134800.1 | - | 4 | 466 | |||
| DA473671.1 | - | 5 | 496 | |||
| GQ502682.1 | - | 2 | 255 | |||
| HQ709264.1 | - | 2 | 234 | |||
| KC677694.1 | - | 2 | 234 | |||
| L36121.1 | - | 2 | 252 | |||
| U35735.1 | - | 4 | 402 | |||
| Y19039.1 | - | 2 | 255 | |||

SLC14A1
Genomic Map