siRNA Details
Gene Symbol: SLC16A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006517.4 | NP_006508.2 | 3 | 1078 | ||
GenBank mRNA | AB085789.1 | - | 3 | 1020 | ||
U05315.1 | - | 3 | 1110 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SLC16A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006517.4 | NP_006508.2 | 3 | 1078 | ||
GenBank mRNA | AB085789.1 | - | 3 | 1020 | ||
U05315.1 | - | 3 | 1110 |