siRNA Details
| Gene Symbol: SMARCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003076.4 | NP_003067.3 | 8, 9 | 1212 | ||
| NM_139071.2 | NP_620710.2 | 8, 9 | 1212 | |||
| XM_005269107.3 | XP_005269164.2 | 7, 8 | 1089 | |||
| GenBank mRNA | AF109733.1 | - | 8, 9 | 1000 | ||
| AK130517.1 | - | 9, 10 | 952 | |||
| AK292700.1 | - | 8, 9 | 876 | |||
| BC009368.2 | - | 8, 9 | 1162 | 4110499 | ||
| DA825057.1 | - | 4, 5 | 483 | |||

SMARCD1
Genomic Map