siRNA Details
| Gene Symbol: SMARCD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098426.1 | NP_001091896.1 | 8 | 1376 | ||
| XM_005257604.2 | XP_005257661.2 | 8 | 923 | |||
| XM_017024967.1 | XP_016880456.1 | 8 | 2115 | |||
| GenBank mRNA | AK300939.1 | - | 8 | 1160 | ||
| AK303951.1 | - | 8 | 962 | |||
| AK307944.1 | - | 8 | 929 | |||
| BC136322.1 | - | 9 | 1245 | 9020309 | ||
| BC142963.1 | - | 8 | 1325 | 8860380 | ||
| BM463728.1 | - | 8 | 873 | |||
| U66618.1 | - | 8 | 1319 | |||

SMARCD2
Genomic Map