siRNA Details
| Gene Symbol: SNAPC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003082.3 | NP_003073.1 | 9 | 1123 | ||
| GenBank mRNA | AK314933.1 | - | 9 | 1123 | ||
| BC014984.1 | - | 9 | 1031 | 4872279 | ||
| BC019038.2 | - | 9 | 1027 | 4590014 | ||
| U44754.1 | - | 9 | 1031 | |||
| Z47542.1 | - | 9 | 1043 | |||

SNAPC1
Genomic Map