siRNA Details
| Gene Symbol: SNRNP70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301069.1 | NP_001287998.1 | 5, 6 | 826 | ||
| NM_003089.5 | NP_003080.2 | 5, 6 | 826 | |||
| GenBank mRNA | AK096783.1 | - | 5, 6 | 515 | ||
| AK293120.1 | - | 5, 6 | 545 | |||
| AL560281.3 | - | 5, 6 | 514 | |||
| BC001315.1 | - | 5, 6 | 499 | 3454479 | ||
| M22636.1 | - | 5, 6 | 490 | |||
| X06812.1 | - | 3, 4 | 174 | |||
| X06815.1 | - | 5, 6 | 470 | |||
| X84841.1 | - | 4, 5 | 319 | |||

SNRNP70
Genomic Map