siRNA Details
| Gene Symbol: SNTB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021021.3 | NP_066301.1 | 5 | 1450 | ||
| GenBank mRNA | AK026095.1 | - | 7 | 1593 | ||
| AK292655.1 | - | 5 | 1524 | |||
| BC098573.1 | - | 5 | 1410 | 30925462 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SNTB1| Gene Symbol: SNTB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021021.3 | NP_066301.1 | 5 | 1450 | ||
| GenBank mRNA | AK026095.1 | - | 7 | 1593 | ||
| AK292655.1 | - | 5 | 1524 | |||
| BC098573.1 | - | 5 | 1410 | 30925462 | ||